Learn More
Greiner Bio-One Small Volume UV-Star™ 384-well, Flat-Bottom Microplate
Click to view available options
Color:
Black
Unit Size:
Pack of 80
Description
- Saves sample volume
- Standardized footprint
- Allows perfect bottom reading even at low working volumes
Service Plan
No
Warranty
No
Specifications
Specifications
| Format | Standard |
| Array | 16 x 24 |
| Autoclavable | Not Autoclavable |
| Packaging | 10/Pk, 80/Cs. |
| Binding Type | Medium Binding |
| Color | Black |
| Material | Cycloolefin |
| Volume (Metric) Well | 4 to 25 μL |
| No. of Wells | 384 |
| Sterility | Sterile |
| Show More |