Learn More
Brady™ Small Universal Condemnation Kit
Product Code. 11786795 Shop All Brady Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11786795 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11786795 Supplier Brady™ Supplier No. 806189
  • ONLINE EXCLUSIVE PRICE
    512.00€
    442.95€ / Pack of 1
    Save 69.05€
    13% Off
In Stock
Add to basket
Add to basket
The Small Universal Condemnation Kit gives you everything you need to lockout gate valves, ball valves, butterfly valves and other valves.
  • Small adjustable locking kit.
Specifications
Product Type Small Universal Condemnation Kit
Description Small adjustable Lockout Kit (FF)
For Use With (Application) Lockout Tagout
Quantity 1 Ea.