Learn More
Hamilton™ Small Hub Removable Needles
Available in custom lengths and point styles to fit your laboratory needs. Hamilton™ Small Hub Removable Needles are manufactured from stainless steel.
126.00€ - 651.00€
Specifications
Material | Stainless Steel |
---|---|
Product Type | Removable Custom Needle |
For Use With (Equipment) | Small Removable Syringes (100μL and smaller) |
Product Code | Brand | Needle Gauge | Needle Point Style | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Needle Gauge | Needle Point Style | Price | Quantity & Availability | ||||
10595855
|
Hamilton™
7731-02 |
26s G | 5 or AS |
366.00€
Pack of 6 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
||||
10492005
|
Hamilton™
7804-04 |
26s G | 2, 3, or 4 |
205.00€
Pack of 6 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
||||
11530332
|
Hamilton™
7803-05 |
33 G | 2, 3, or 4 |
342.00€
Pack of 6 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
||||
10798204
|
Hamilton™
7804-02 |
22s G | 2, 3, or 4 |
166.00€
Pack of 6 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
||||
11573294
|
Hamilton™
7731-03 |
22 G | 5 or AS |
360.00€
Pack of 6 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
||||
12386439
|
Hamilton™
7804-06 |
18 G | 2, 3, or 4 |
166.00€
Pack of 6 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
||||
11510332
|
Hamilton™
7803-03 |
31 G | 2, 3, or 4 |
341.00€
Pack of 6 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
||||
10698205
|
Hamilton™
7804-03 |
26 G | 2, 3, or 4 |
312.00€
Pack of 6 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
||||
12316299
|
Hamilton™
7731-08 |
23s G | 5 or AS |
366.00€
Pack of 6 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
||||
11500332
|
Hamilton™
7803-02 |
28 G | 2, 3, or 4 |
334.00€
Pack of 6 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
||||
10725873
|
Hamilton™
7731-04 |
22s G | 5 or AS |
360.00€
Pack of 6 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
||||
11532525
|
Hamilton™
7804-01 |
22 G | 2, 3, or 4 |
166.00€
Pack of 6 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
||||
12702257
|
Hamilton™
7803-06 |
29 G | 2, 3, or 4 |
334.00€
Pack of 6 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
||||
10776063
|
Hamilton™
7803-07 |
30 G | 2, 3, or 4 |
651.00€
Pack of 6 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
||||
11520332
|
Hamilton™
7803-04 |
32 G | 2, 3, or 4 |
334.00€
Pack of 6 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
||||
10140888
|
Hamilton™
7731-01 |
26 G | 5 or AS |
366.00€
Pack of 6 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
||||
15927335
|
Hamilton™
7804-03 |
26 G | 2, 3, or 4 |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
Description
- Autoclavable
- Standard needle length is 2"/51mm
- Can be used with (RN)-style syringes that are 100μL and smaller
Spécification
Stainless Steel | |
Removable Custom Needle | |
Small Removable Syringes (100μL and smaller) |