Learn More
Asecos™ SLX-CLASSIC Recirculating Air Safety Storage Cabinets
GREENER_CHOICE
Product Code. 15527839 Shop All Asecos Products
Change view
Click to view available options
Load Bearing Capacity:
12 x Shelves with sump: 25 kg/11 L each, Maximum load: 300 kg
6 x Shelves with sump: 25 kg/11 L each, Maximum load: 300 kg
8 x Shelves with sump 25 kg/11 L each, Maximum load 300 kg
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Load Bearing Capacity unitSize
15527839 8 x Shelves with sump 25 kg/11 L each, Maximum load 300 kg Each
15695677 12 x Shelves with sump: 25 kg/11 L each, Maximum load: 300 kg Each
15904690 6 x Shelves with sump: 25 kg/11 L each, Maximum load: 300 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15527839 Supplier Asecos™ Supplier No. 3293600130607
For the safe and approved storage of aggressive, non-flammable hazardous materials in workrooms.
  • Robust construction and longevity – cabinet body made of powder-coated sheet steel with safety elements assembled outside the storage compartment for increased protection against corrosion
  • Two-in-one – two hermetically separated storage compartments allow the isolated storage of acids and bases in one cabinet
  • No unauthorized use – doors lockable with profile cylinder with integration in an existing locking system possible
  • Easy alignment – adjusting aids to compensate for uneven floors
  • Recirculating air filter system, ideal for the direct installation in working areas with very low noise, only approx. 39 dB
  • With multi-stage wideband filter
  • Monitoring electronics for extraction include cool-device cable and power plug
  • Optical and acoustic alarm including potential-free alarm contact
  • Display with innovative touch panel
  • Manual check of the filter saturation
  • Eco-friendly with longer filter lifetimes and a reusable filter system
Specifications
Certifications/Compliance EN 14727
Configuration 8 x 25 kg/11 L PP pull-out shelves with PP sumps
Dimensions (D x W x H) Interior 565 x 485 x 1865 mm
Dimensions (D x W x H) 615 x 1197 x 2300 mm
Door Style Wing
For Use With (Application) Storage of aggressive, non-flammable hazardous materials
Load Bearing Capacity 8 x Shelves with sump 25 kg/11 L each, Maximum load 300 kg
No. of Doors 2
No. of Shelves 8
Shelf Type Pull-out shelf
Surface Coating Powder-coated
Weight (Metric) 330.7 kg
Color Light Gray
Depth (English) Exterior 24.21 in.
Height (English) Exterior 90.55 in.
Height (Metric) Opening 1865 mm
Material Steel
Product Type Safety Storage Cabinet
Width (English) Exterior 47.13 in.