missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Safety-Walk™ Slip Resistant Coarse Tape, Series 700
Description
- Anti-slip tape for extreme and heavy traffic areas
- Extra-large abrasive particles resist clogging in muddy or oily conditions
- Can be painted without losing friction properties
- High performance adhesive
- Suitable for use on agriculture and forestry equipment, road building, civil engineering, lorries, tractors, construction vehicles, cranes, and building equipment
Specifications
Specifications
| Certifications/Compliance | DIN 51130: R 13, EN 13501 1: Bfl s1 |
| Description | Extra Strong (Type 4) |
| Dimensions (L x W) | 36.6 x 0.31 m |
| For Use With (Application) | Additional Slip Protection in Extreme, Heavily Frequented Areas |
| Packaging Quantity | 1 Roll |
| Adhesive Type | Pressure Sensitive Adhesive |
| Color | Black |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?