Learn More
Thermo Scientific™ Racks for Thermo Scientific™ 5 Shelf TSX/TDE Series Freezers
Shop All Thermo Scientific ProductsDescripción
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for TSX/TDE 5 shelf Series Freezers as well as well as previous generation Revco™ ExF, DxF and HERAfeeze™ HFU B Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
Thermo Scientific™ freezer racking and tubing solutions offer a convenient, streamlined process to maximize sample storage in your laboratory. Our racking solutions are designed to hold tubes ranging from 200μL up to 5.0mL, offering maximized sample storage capacity for both five and four inner door freezers.
Especificaciones
Especificaciones
| Product Type | Sliding Drawer Freezer Rack (Tube Boxes) |
| Description | Front Access Drawer Rack |
| Color | Silver |
| Holds | Matrix 2D Tubes (500 μL) |
| Material | Stainless Steel |
| Positions | 28 Boxes/Rack |
| Shape | Rectangular |
| For Use With (Equipment) | EXF600, HFU600B (28 cu.ft.) |
| Freezer Compatiblity (Inner Doors) | 5 |
| No. of Racks | 6 |
| Mostrar más |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.