Learn More
Thermo Scientific™ Racks for Thermo Scientific™ 5 Shelf TSX/TDE Series Freezers
Product Code. 11319321 Shop All Thermo Scientific Products
Click to view available options
Holds:
Boxes (2 in.)
Boxes (3 in.)
Matrix 2D Internally Threaded Tubes (200 μL)
Matrix 2D Tubes (1 mL)
Matrix 2D Tubes (500 μL)
Microplates
Microplates with Lids
Nunc or Matrix 2D Tubes (5 mL)
Tubes
Positions:
12 Boxes/Rack
16 Boxes/Rack
16 Plates/Rack
18 Boxes/Rack
21 Boxes/Rack
24 Boxes/Rack
24 Plates/Rack
28 Boxes/Rack
28 Plates/Rack
30 Plates/Rack
36 Boxes/Rack
42 Boxes/Rack
44 Plates/Rack
48/Box
66 Plates/Rack
70 Plates/Rack
84 Plates/Rack
9 Boxes/Rack
96/Box
For Use With (Equipment):
EXF240, HFU240B (13 cu.ft.)
EXF240, HFU240B (5 shelf) (13 cu.ft.)
EXF320, EXF400, HFU320B, HFU400B (17.3, 23 cu.ft.)
EXF320, EXF400, HFU320B, HFU400B (5 shelf) (17.3, 23 cu.ft.)
EXF600, HFU600B (28 cu.ft.)
Revco UxF, HERAFreeze HFU-T (14.9, 19.4, 24.1, 28.8, 33.5 cu.ft.)
Upright Freezers (13 cu.ft.)
Upright Freezers (17.3, 23 cu.ft.)
Upright Freezers (17.3, 23, 28.8 cu.ft.)
Unit Size:
Each
Product Code. 11319321
Brand: Thermo Scientific™ 398326
Maximize tube storage capacity per freezer with a racking solution for five inner-door, ultra-low temperature freezers.
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for TSX/TDE 5 shelf Series Freezers as well as well as previous generation Revco™ ExF, DxF and HERAfeeze™ HFU B Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
  • Warranty varies by country, please contact us for details
TRUSTED_SUSTAINABILITY
Specifications
Product Type Side Access Freezer Rack (Adjustable)
Description 2 in. Box Racks - Adjustable side access rack
Color Silver
Holds Boxes (2 in.)
Material Stainless Steel
Positions 16 Boxes/Rack
Shelves 4 Tiers
For Use With (Equipment) Upright Freezers (17.3, 23 cu.ft.)
Width (Metric) 13.7 cm
Width (English) 5.4 in.
Height (Metric) 23.9 cm
Height (English) 9.4 in.
Depth (English) 22.1 in.
Depth (Metric) 56.1 cm