Learn More
- Home
- Products
- Racks
- Freezer Storage Racks
- Thermo Scientific™ Racks for Thermo Scientific™ 4 Shelf TSX/TDE Series Freezers
Thermo Scientific™ Racks for Thermo Scientific™ 4 Shelf TSX/TDE Series Freezers
Product Code. p-6020011 Shop All Thermo Scientific Products| Product Code. | Holds | Product Type | For Use With (Equipment) | unitSize |
|---|---|---|---|---|
| 15150529 | Matrix Internally Threaded Tubes (1 mL) | Sliding Drawer Freezer Rack (Tube Boxes) | Forma 88000 and TSU Series | Each |
| 15140519 | Tubes, 32 Boxes/Rack | Sliding Drawer Freezer Rack (Tubes) | 900 Series 13CF, TSE240 | Each |
| 15150519 | Tubes, 48 Boxes/Rack | Sliding Drawer Freezer Rack (Tubes) | 900 Series 17-23CF, TSE320, TSE400 | Each |
| 15170519 | Tubes, 56 Boxes/Rack | Sliding Drawer Freezer Rack (Tubes) | 900 Series 28CF, TSE600 | Each |
| 15160519 | Matrix 2D Tubes (500 μL) | Sliding Drawer Freezer Rack (Tube Boxes) | Forma 88000 and TSU Series | Each |
| 15180519 | Tubes, 24 Boxes/Rack | Sliding Drawer Freezer Rack (Tubes) | 900 Series 13CF, TSE240 | Each |
| 15190519 | Nunc Cryobank Internally Threaded Tubes or Matrix Screw Top Tubes (500 μL) | Sliding Drawer Freezer Rack (Tube Boxes) | 900 Series 17-23CF, TSE320, TSE400 | Each |
| 15110529 | Nunc Cryobank Internally Threaded Tubes or Matrix Screw Top Tubes (500 μL) | Sliding Drawer Freezer Rack (Tube Boxes) | 900 Series 28CF, TSE600 | Each |
| 15100529 | Nunc Cryobank Internally Threaded Tubes or Matrix Screw Top Tubes (500 μL) | Sliding Drawer Freezer Rack (Tube Boxes) | Forma 88000 and TSU Series | Each |
| 15120529 | Tubes, 16 Boxes/Rack | Sliding Drawer Freezer Rack (Tubes) | 900 Series 13CF, TSE240 | Each |
| 15130529 | Matrix Internally Threaded Tubes (1 mL) | Sliding Drawer Freezer Rack (Tube Boxes) | 900 Series 17-23CF, TSE320, TSE400 | Each |
| 15140529 | Tubes, 28 Boxes/Rack | Sliding Drawer Freezer Rack (Tubes) | 900 Series 28CF, TSE600 | Each |
| 15100539 | Microplates, 20 Boxes/Rack | Sliding Drawer Freezer Rack (Tubes) | 900 Series 13CF, TSE240 | Each |
| 15110539 | Microplates | Sliding Drawer Freezer Rack (Tube Boxes) | 900 Series 17-23CF, TSE320, TSE400 | Each |
| 15120539 | Tubes (1.0 to 1.4 mL or 1.8 mL), 35 Boxes/Rack | Sliding Drawer Freezer Rack (Tubes) | 900 Series 28CF, TSE600 | Each |
| 11810553 | Externally Threaded Tubes (1.8 mL) | Sliding Drawer Freezer Rack (Tube Boxes) | Forma 88000 and TSU Series | Each |
| 15170529 | Tubes, 18 Boxes/Rack | Sliding Drawer Freezer Rack (Tubes) | EXF and HFU 200, 300, 400 and 600 Series | Each |
| 15180529 | Tubes, 21 Boxes/Rack | Sliding Drawer Freezer Rack (Tubes) | EXF and HFU 200, 300, 400 and 600 Series | Each |
| 15190529 | Nunc Cryobank Internally Threaded Tubes (5 mL) | Sliding Drawer Freezer Rack (Tube Boxes) | Revco UxF, HERAFreeze HFU-T Series | Each |
| 13114544 | Boxes (2 in.) | Sliding Drawer Rack (Tube Boxes) | Forma 900, Forma 7000, TSE, TSD Sereis (28 cu.ft.) | Each |
| 11870543 | Boxes (2 in.) | Side Access Freezer Rack (Adjustable) | TDE and TSX Series (4 shelves) | Each |
| 11880543 | Boxes (3 in.) | Side Access Freezer Rack (Adjustable) | TDE and TSX Series (4 shelves) | Each |
| 11890543 | Microplates | Side Access Freezer Rack (Microplate/Deepwell) | TDE and TSX Series (4 shelves) | Each |
| 11860543 | Microplates | Side Access Freezer Rack (Microplate/Deepwell) | TDE and TSX Series (4 shelves) | Each |
| 10744984 | Boxes (2 in.) | Sliding Drawer Rack (Tube Boxes) | TDE and TSX Series (4 shelves) | Each |
| 13167444 | Boxes (3 in.) | Sliding Drawer Rack (Tube Boxes) | TDE and TSX Series (4 shelves) | Each |
| 11706469 | Boxes (3 in.) | Sliding Drawer Rack (Tube Boxes) | TDE and TSX Series (4 shelves) | Each |
Descripción
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for TSX/TDE 4 shelf Series Freezers as well as well as previous generation Forma™ 88000 and TSU Series Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
Thermo Scientific freezer racking and tubing solutions offer a convenient, streamlined process to maximize sample storage in your laboratory. Our racking solutions are designed to hold tubes ranging from 200μL up to 5.0mL, offering maximized sample storage capacity for both five and four inner door freezers.
Especificaciones
Especificaciones
| Description | Front Access Drawer Rack |
| Color | Silver |
| Holds | Matrix Internally Threaded Tubes (1 mL) |
| Material | Stainless Steel |
| Positions | 28 Boxes/Rack |
| For Use With (Equipment) | Forma 88000 and TSU Series |
| Freezer Compatiblity (Inner Doors) | 4 |
| No. of Racks | 3 (300 capacity) 4 (400 capacity) 5 (500 capacity) 6 (600 capacity) 7 (700 capacity) |
| No. of Racks per Freezer | 12 (300 capacity) 16 (400 capacity) 20 (500 capacity) 24 (600 capacity) 28 (700 capacity) |
| Width (English) | 11.9 in. |
| Height (English) | 26.96 in. |
| Depth (English) | 5.5 in. |
| Depth (Metric) | 14 cm |
| Product Type | Sliding Drawer Freezer Rack (Tube Boxes) |
| Shelves | 6 |
| Height (Metric) | 66.5 cm |
| Width (Metric) | 30.2 cm |
| Mostrar más Mostrar menos |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.