Learn More
BRAND™ Slide Staining Trough
Product Code. 10251951 Shop All Brand Products
Change view
Click to view available options
Material:
POM
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
10251951 POM Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10251951 Supplier BRAND™ Supplier No. 471500
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Slide Staining Trough with a close fitting lid that helps protect against evaporation.
  • For 25 slides
  • Close fitting lid helps protect against evaporation
  • Tray has a fold down handle
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Slides
Type Staining Trough
Material POM