Learn More
Hettich Lab Technology™ Slide Carrier
Click to view available options
Capacity:
1, 2, 4, 8 mL, 3 x 2 mL, 4 x 1 mL
2 x 1, 2, 4, 8 mL, 3 x 2 mL, 4 x 1 mL
Unit Size:
Each
Description
- For cyto centrifuge systems and cell evaluations
- Accepts funneling cyto chambers 1663-1668
Specifications
Specifications
| Type | Slide Carrier |
| For Use With (Equipment) | Cyto centrifuge system, Filter cards 1691-1694, Funneling cyto chambers 1663-1668 |
| Capacity | 2 x 1, 2, 4, 8 mL, 3 x 2 mL, 4 x 1 mL |
| Autoclavable | Autoclavable |
| Includes | 2 Adapter Rings |