Learn More
Hettich Lab Technology™ Slide Carrier
Product Code. 10352402
Click to view available options
Capacity:
1, 2, 4, 8 mL, 3 x 2 mL, 4 x 1 mL
2 x 1, 2, 4, 8 mL, 3 x 2 mL, 4 x 1 mL
Unit Size:
Each
Product Code. 10352402
Brand: Hettich Lab Technology™ 1662
Hettich Lab Technology™ Slide Carrier is available as a single or double chamber slide/filter carrier for cyto-centrifuge systems.
  • For cyto centrifuge systems and cell evaluations
  • Accepts funneling cyto chambers 1663-1668
Specifications
Type Slide Carrier
For Use With (Equipment) Cyto centrifuge system, Filter cards 1675-1678, Funneling cyto chambers 1663-1668
Capacity 1, 2, 4, 8 mL, 3 x 2 mL, 4 x 1 mL
Autoclavable Autoclavable
Includes Adapter ring