Learn More
Karl Hecht™ Assistent™ Antistatic Slide Cases
Provides for secure organization, storage and transport for laboratory slides. Karl Hecht™ Assistent™ Antistatic Slide Cases have a frame manufactured from polypropylene plastic.
17.81€ - 21.25€
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
10023721
|
Karl Hecht™
42701010 |
100 microslides |
21.25€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
10195861
|
Karl Hecht™
42702010 |
50 microslides |
17.81€
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
Description
- Stackable
- Black in color
- Resistant to xylene
- With numbered plastic grooves
Specifications
Slide Box | |
1 Ea. |