Learn More
Karl Hecht™ Assistent™ Antistatic Slide Cases
Provides for secure organization, storage and transport for laboratory slides. Karl Hecht™ Assistent™ Antistatic Slide Cases have a frame manufactured from polypropylene plastic.
Brand: Karl Hecht™ 42701010
Additional Details : Weight : 0.30000kg
Description
- Stackable
- Black in color
- Resistant to xylene
- With numbered plastic grooves
Specifications
Slide Box | |
100 microslides | |
1 Ea. |