missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC24A6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Specifications
Antigen | SLC24A6 |
---|---|
Concentration | 0.25 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16871025
|
Proteintech
21430-1-AP-20UL |
20 μL |
140.00€
20µL |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
16861025
|
Proteintech
21430-1-AP-150UL |
150 μL |
399.00€
150µL |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
Description
SLC24A6 belongs to a family of potassium-dependent sodium/calcium exchangers that maintain cellular calcium homeostasis through the electrogenic counter transport of 4 sodium ions for 1 calcium ion and 1 potassium ion.Specifications
SLC24A6 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q6J4K2 | |
80024 | |
SLC24A6 Fusion Protein Ag16034 | |
Primary | |
-20°C | |
SLC8B1 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
NCKX6, NCLX, SLC24A6 | |
SLC8B1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |