Learn More
Bollé Safety Slam Safety Glasses
Click to view available options
Lens Tint:
Shade 1.7
Shade 3
Shade 5
Yellow
Lens Coating:
Anti-fog, Impact-resistant, Scratch-resistant
Impact-resistant, Scratch-resistant
Unit Size:
Each
Description
- Stylish.
- Easy to wear.
- Protection at all times.
- Unbeatable comfort.
- EN 166: Guaranteed minimum resistance of the protective eyewear against everyday risks including dropping the protective eyewear onto the ground, ageing by light exposure, exposure to heat or corrosion etc.
- EN 170, Risks relating to radiation - UV protection: The protection filters ultraviolet radiation. This type of protection is particularly recommended during activities where the source of ultraviolet radiation is predominant, such as mercury vapor lamps, actinic or germicides. Levels of application: 2-1.2, 2C-1.2 Lens color: Clear, blue, yellow or green. Adding the code 'C' indicates good color recognition by the holder.
- Mechanical risk F - Mechanical risk - Low energy impact:& Low energy impact, resists a 6 mm, 0.86 g ball at 45 m/s.
- Mechanical risk T - High speed particles at extreme temperatures: The letter T, immediately after the mechanical strength symbol, authorizes use for high-speed particles at extreme temperatures.
- Optical class: Class 1, continuous work, worn permanently.
- CE, European standard: Frame marked to indicate meeting CE standard.
Specifications
Specifications
| Lens Coating | Impact-resistant, Scratch-resistant |
| Description | Model SLAWPCC2 |
| Certifications/Compliance (Europe) | EN166, EN170, EN172, EN175/1.7, Mechanical risk F/T, Optical class 1, CE |
| Lens Tint | Shade 1.7 |
| Product Type | Safety Glasses |
| For Use With (Application) | Welding, protection, continuous wear |
| No. per Case | 200/Cs. |
| No. per Pack | 1/bag, 10/Pk. |
| Weight (Metric) | 24 g |
| Lens Material | Polycarbonate |
| Show More |