missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLAM/CD150 Mouse anti-Human, Alexa Fluor 647, Clone: 542301, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB1642R-100UG
This item is not returnable.
View return policy
Description
SLAM/CD150 Monoclonal specifically detects SLAM/CD150 in Human samples. It is validated for Flow Cytometry.Specifications
| SLAM/CD150 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q13291 | |
| SLAMF1 | |
| Mouse myeloma cell line NS0-derived recombinant human SLAM/CD150 Ala21-Pro237 Accession # Q13291 | |
| RUO | |
| 6504 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 542301 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD 150, CD150, CD150IPO-3, CDw150, IPO-3, signaling lymphocytic activation molecule, signaling lymphocytic activation molecule family member 1, SLAMCD150 antigen, SLAMF1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human SLAM/CD150 in direct ELISAs and Western blots. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction