Description
- Wrap-around fit
- All-round vision
- Comfortable straight temples
Specifications
Specifications
| Certifications/Compliance | Risks relating to radiations - UV Protection - EN 170 - (2-1.2, 2C-1.2); Mechanical risk - Low energy impact - F; Mechanical risk- High speed particles at extreme temperatures - T |
| Lens Coating | Anti-fog, Scratch-resistant |
| Description | Tinted |
| Frame Color | Black |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Informativa sulla privacy.
Individuate un'opportunità di miglioramento?