Learn More
Asecos™ SL-CLASSIC-UB Underbench Safety Storage Cabinets with 3 Doors
Product Code. 15955770 Shop All Asecos Products
Change view
Click to view available options
Load Bearing Capacity:
2 x Pull-out shelves: 25 kg/11 L each, Maximum load: 100 kg
4 x Pull-out shelves: 25 kg/11 L each, Maximum load: 100 kg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Load Bearing Capacity Unit Size
15955770 4 x Pull-out shelves: 25 kg/11 L each, Maximum load: 100 kg Each
15874178 2 x Pull-out shelves: 25 kg/11 L each, Maximum load: 100 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15955770 Supplier Asecos™ Supplier No. 3061689230618
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
For the safe and proper storage of aggressive, non-flammable hazardous materials in working areas underneath work benches and fume cupboards.
  • Robust construction and longevity – underbench cabinets completely made of melamine resin and flame retardant special plates
  • Safe and separate storage – acids and alkalis can be stored separately
  • Two hermetically divided storage compartments, integrated aeration and venting system, corrosion resistance of the metal-free air ducts
  • In accordance with TRGS 510
  • Adjusting aids are integrated in the cabinet base, fast and effective compensation for uneven floors
  • Interior equipment available includes pull-out shelves and bottom collecting sumps
  • No unauthorized use – doors lockable with profile cylinder (integration into an existing locking system possible)
  • Ventilation – integrated metal-free air ducts ready for connection to a technical exhaust system with only one extraction air inlet
Specifications
Certifications/Compliance TRGS 510
Configuration 4 x 25 kg/11 L PP pull-out shelves with PP sump
Dimensions (D x W x H) Interior 531 x 489 x 562 mm
Dimensions (D x W x H) 570 x 1400 x 600 mm
Door Style Wing
For Use With (Application) Storage of aggressive and non-flammable hazardous substances
Load Bearing Capacity 4 x Pull-out shelves: 25 kg/11 L each, Maximum load: 100 kg
No. of Doors 3
No. of Shelves None
Weight (Metric) 103.5 kg
Color Light Gray
Depth (English) Exterior 22.44 in.
Height (English) Exterior 23.62 in.
Material Melamine Resin
Product Type Safety Storage Cabinet
Width (English) Exterior 55.12 in.