Learn More
Asecos™ SL-CLASSIC Safety Storage Cabinets
Product Code. 15600391 Shop All Asecos Products
Change view
Click to view available options
Door Style:
Wing
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Door Style Unit Size
15600391 Wing Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15600391 Supplier Asecos™ Supplier No. 3061300130615
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
For the safe and proper storage of aggressive, non-flammable hazardous materials in working areas.
  • Robust construction and longevity – tall cabinet with body made of powder-coated sheet steel and interior space made of melamine resin and flame retardant special plates
  • Safe and separate storage – acids and alkalis can be stored separately
  • Two hermetically divided storage compartments, integrated aeration and venting system, corrosion resistance of the metal-free air ducts
  • In accordance with TRGS 510
  • Adjusting aids are integrated in the cabinet base, fast and effective compensation for uneven floors
  • Interior equipment available includes pull-out shelves and bottom collecting sumps
  • No unauthorized use – doors lockable with profile cylinder (integration into an existing locking system possible)
  • Ventilation – integrated metal-free air ducts ready for connection to a technical exhaust system with only one extraction air inlet
Specifications
Certifications/Compliance TRGS 510
Configuration 1 x light gray wing door; 6 x PP pull-out shelves with PP sump, load capacity 25 kg, capacity 11 L
Dimensions (D x W x H) Interior 570 x 485 x 914 mm
Dimensions (D x W x H) 603 x 597 x 1965 mm
Door Style Wing
For Use With (Application) Storage of aggressive, non-flammable hazardous substances
Load Bearing Capacity 300 kg
No. of Doors 1
No. of Shelves None
Surface Coating Powder-coated
Weight (Metric) 176.4 kg
Color Light Gray
Depth (English) Exterior 23.74 in.
Height (English) Exterior 77.36 in.
Material Steel
Product Type Safety Storage Cabinet
Width (English) Exterior 23.50 in.