missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ SL 327
Description
Selective inhibitor of MEK1 and MEK2 (IC50 values are 0.18 and 0.22 μM respectively); blocks hippocampal LTP in vitro. Brain penetrant in vivo, blocking fear conditioning and learning in rats, and producing neuroprotection in mice, following systemic administration.
Specifications
Specifications
| Quantity | 1 mg |
| Product Line | Tocris |
| Formula Weight | 335.35 |
| Purity | >99% |
| Molecular Weight (g/mol) | 335.348 |
| Chemical Name or Material | SL 327 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?