missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SJFd
Description
SJFδ is a potent and selective PROTAC ™ Degrader of p38δ (DC50 = 46.17 nM; Dmax = 99.41%). SJFδ comprises the multikinase inhibitor foretinib joined by a linker to a VHL ligand. Exhibits no significant degradation of p38α, β or γ. Selectively degrades p38δ in MDA-MB-231 cells. PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | N-(3-Fluoro-4-((7-(2-(2-(2-(2-(((2S,4R)-4-hydroxy-1-((S)-3-methyl-2-(1-oxoisoindolin-2-yl)butanoyl)pyrrolidine-2-carboxamido)methyl)-5-(4-methylthiazol-5-yl)phenoxy)ethoxy)ethoxy)ethoxy)-6-methoxyquinolin-4-yl)oxy)phenyl)-N-(4-fluorophenyl)cyclopropane-1, |
| CAS | 2254609-23-7 |
| Quantity | 5 mg |
| Target | Active Degraders |
| Molecular Formula | C62H63F2N7O12S |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?