Learn More
Axygen™ Single Well High Profile Reagent Reservoirs
Shop All Axygen Scientific Products
Click to view available options
Capacity (Metric):
240 mL
282 mL
290 mL
Sterility:
Non-sterile
Sterile
Channels:
8
12
96
384
Unit Size:
Pack of 25
Description
Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.
- Durable and cost effective
- Various formats for multiple applications
Specifications
Specifications
| Certifications/Compliance | ANSI/SLAS, RNase/DNase-free, non-pyrogenic |
| Color | Clear |
| Capacity (Metric) | 240 mL |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Type | Reagent Reservoir |
| No. per Case | 25/Cs. |
| No. per Pack | 25/Pk. |
| Packaging | 25/Cs. |
| Channels | 96 |
| Show More |