Learn More
Tenak™ Single Tray Rack for Upright Freezers, Standard
Product Code. 12446980 Shop All TENAK Products
Click to view available options
Dimensions (L x W x H):
420 x 139 x 170 mm
420 x 139 x 280 mm
560 x 139 x 170 mm
560 x 139 x 385 mm
Unit Size:
Each
Product Code. 12446980
Brand: Tenak™ TE29120
Trays designed for applications where the samples are either not stored in boxes or has odd sizes.
  • Single trays are a drawer including a lid with a handle on the front and it will fit into any freezer
  • Racks can be used with all freezer brands
  • Constructed of stainless steel
Specifications
Material Stainless Steel
For Use With (Equipment) Upright Freezers
Dimensions (L x W x H) 560 x 139 x 170 mm
Width (Metric) 139 mm