missing translation for 'onlineSavingsMsg'
Learn More

Simvastatin Hydroxy Acid Methyl Ester, TRC

Product Code. 30132737
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132737 100 mg 100mg
30004332 10 mg 10mg
30010521 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132737 Supplier Toronto Research Chemicals Supplier No. TRCS485015100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145576-26-7
Molecular Formula C26H42O6
Molecular Weight (g/mol) 450.61
Synonym 1-Naphthaleneheptanoic acid, 8-(2,2-dimethyl-1-oxobutoxy)-1,2,6,7,8,8a-hexahydro-β,δ-dihydroxy-2,6-dimethyl-, methyl ester, (βR,δR,1S,2S,6R,8S,8aR)-, 1-Naphthaleneheptanoic acid, 8-(2,2-dimethyl-1-oxobutoxy)-1,2,6,7,8,8a-hexahydro-β,δ-dihydroxy-2,6-dimethyl-, methyl ester, [1S-[1α(βS*,δS*),2α,6β,8β,8aα]]-, Methyl (βR,δR,1S,2S,6R,8S,8aR)-8-(2,2-dimethyl-1-oxobutoxy)-1,2,6,7,8,8a-hexahydro-β,δ-dihydroxy-2,6-dimethyl-1-naphthaleneheptanoate, Simvastatin Hydroxy Acid Methyl Ester
IUPAC Name methyl (3R,5R)-7-[(1S,2S,6R,8S,8aR)-8-(2,2-dimethylbutanoyloxy)-2,6-dimethyl-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate
SMILES CCC(C)(C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC(=O)OC)[C@@H]12

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C26H42O6/c1-7-26(4,5)25(30)32-22-13-16(2)12-18-9-8-17(3)21(24(18)22)11-10-19(27)14-20(28)15-23(29)31-6/h8-9,12,16-17,19-22,24,27-28H,7,10-11,13-15H2,1-6H3/t16-,17-,19+,20+,21-,22-,24-/m0/s1
Formula Weight 450.2981
Chemical Name or Material Simvastatin Hydroxy Acid Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.