missing translation for 'onlineSavingsMsg'
Learn More

Simvastatin 4’-Methyl Ether, TRC

Product Code. 30412334
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412334 50 mg -
30414212 5 mg 5mg
30413387 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30412334 Supplier Toronto Research Chemicals Supplier No. TRCS48504550MG
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 864357-88-0
Molecular Formula C26 H40 O5
Molecular Weight (g/mol) 432.59
Synonym Butanoic acid, 2,2-dimethyl-, (1S,3R,7S,8S,8aR)-1,2,3,7,8,8a-hexahydro-3,7-dimethyl-8-[2-[(2R,4R)-tetrahydro-4-methoxy-6-oxo-2H-pyran-2-yl]ethyl]-1-naphthalenyl ester, (1S,3R,7S,8S,8aR)-1,2,3,7,8,8a-Hexahydro-3,7-dimethyl-8-[2-[(2R,4R)-tetrahydro-4-methoxy-6-oxo-2H-pyran-2-yl]ethyl]-1-naphthalenyl 2,2-dimethylbutanoate, (1S,3R,7S,8S,8aR)-8-[2-[(2R,4R)-4-Methoxy-6-oxooxan-2-yl]ethyl]-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl 2,2-dimethylbutanoate
IUPAC Name [(1S,3R,7S,8S,8aR)-8-[2-[(2R,4R)-4-methoxy-6-oxooxan-2-yl]ethyl]-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl] 2,2-dimethylbutanoate
SMILES CCC(C)(C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@H](CC(=O)O3)OC)[C@@H]12

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C26H40O5/c1-7-26(4,5)25(28)31-22-13-16(2)12-18-9-8-17(3)21(24(18)22)11-10-19-14-20(29-6)15-23(27)30-19/h8-9,12,16-17,19-22,24H,7,10-11,13-15H2,1-6H3/t16-,17-,19+,20+,21-,22-,24-/m0/s1
Formula Weight 432.2876
Chemical Name or Material Simvastatin 4'-Methyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.