Learn More
Vitlab™ Simplex II Fix Dispenser
Product Code. p-7146509
Click to view available options
Volume (Metric):
1 mL
5 mL
10 mL
Unit Size:
Each
Product Code. 15610925
Brand: Vitlab™ 1622502
Vitlab™ Simplex II Fix Dispenser is designed to be practically universally applicable and can be used with many organic and inorganic solutions.
  • Bottle-top dispenser with fixed volume
  • Direct displacement plunger with broad PFA seal prevents crystallisation build-up
  • Screwable ventilation plug for easy connection of a drying tube
  • Screwable discharge valve is equipped with an additional safety bulb and closes the dosing channel, if dispensing tube is not installed, so that no medium can escape
  • Equipped with a positive displacement piston and a fluoroplastic (PFA) sealing lip on the cylinder wall
  • Nominal volume 1 to 10mL includes adapters GL 25, GL 28, GL 32, GL 38, S 40 and telescopic intake tube (length 125 to 240mm)
Specifications
Volume (Metric) 1 mL