missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ Simplex™ Dispenser

Product Code. 10100312
Change view
Click to view available options
Volume (Metric):
0.1 to 2.5 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10100312 0.1 to 2.5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10100312 Supplier VITLAB™ Supplier No. 1601503

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Accuracy ≤±0.6%
Product Type Bottletop Dispenser
Certifications/Compliance DIN EN 285, DE-M marked
Volume (Metric) 0.1 to 2.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.