Learn More
Witeg™ Simple Distillation Kit
Click to view available options
Capacity (Metric):
250mL
Unit Size:
Each
Description
- Ideal for easy, smaller scale distllation processes
- Includes boiling flask, column, condenser, spigot, clamps
Specifications
Specifications
| Includes | Boiling flask, column, condenser tube, spigot, clamps |
| Type | Distillation Kit |
| Capacity (Metric) | 250mL |