Learn More
Silver nitrate, ACS Reagent, ≥99.0%, Honeywell Fluka™
Product Code. 15611890 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
Poly bottle
Unit Size:
100g
25g
500g
Product Code. 15611890
Brand: Honeywell-Fluka 209139500G
Request Bulk or Custom Format
ACS Reagent,≥99.0%
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
SMILES [Ag+].[O-][N+]([O-])=O
Specifications
Quantity 500 g
Linear Formula AgNO3
UN Number 1493
IUPAC Name silver(1+) nitrate
Formula Weight 169.87g/mol
Percent Purity ≥99%
Grade ACS Reagent
Packaging Poly bottle
Density Vapor Density: 5.8 (vs air)
Chemical Name or Material Silver Nitrate