Learn More
BRAND™ Silicone Tubing
Product Code. 10674162 Shop All Brand Products
Click to view available options
Diameter (Metric) Outer:
4 mm
5 mm
6 mm
9 mm
12 mm
Unit Size:
25m
Product Code. 10674162
Brand: BRAND™ 143359
Translucent tubing
  • Autoclavable (121 °C)
  • Note: limited chemical resistance when used with concentrated acids
  • TRUSTED_SUSTAINABILITY
    Specifications
    Diameter (Metric) Inner 6 mm
    Material Silicone Rubber
    Diameter (Metric) Outer 9 mm
    Overall Length (Metric) 25 m
    Tubing Size 6 x 9 mm
    Color Translucent