Learn More
Fisherbrand™ Silicone Tubing
Product Code. 11527083 Shop All Fisherbrand Products
Click to view available options
Diameter (Metric) Inner:
1.0 mm
2 mm
3 mm
4 mm
5 mm
6 mm
6.5 mm
8 mm
9 mm
10.0 mm
12.5 mm
16 mm
Thickness (Metric):
0.75 mm
1.0 mm
1.5 mm
2 mm
2.0 mm
2.25 mm
25 mm
Unit Size:
10m
Product Code. 11527083
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For consistently reliable and accurate fluid transfers.
Available in different diameters
Specifications
Diameter (Metric) Inner 8 mm
Overall Length (Metric) 10 m
Thickness (Metric) 2 mm