Learn More
BRAND™ Silicone Stoppers
Product Code. 10243201 Shop All Brand Products
Click to view available options
Diameter (Metric) Bottom:
12.5 mm
14 mm
17 mm
21 mm
Unit Size:
Pack of 10
Product Code. 10243201
Brand: BRAND™ 144340
Silicone construction with natural color. Prevents spillage while working in the lab.
  • Silicone rubber construction
  • Used to prevent inadvertant leaks and spills
  • Natural color
TRUSTED_SUSTAINABILITY
Specifications
Height (Metric) 30 mm
Quantity 10
Color Natural
Diameter (Metric) Bottom 21 mm
Diameter (Metric) Top 27 mm
Material Silicone