Learn More
BRAND™ Silicone Stopper
Product Code. p-7164805 Shop All Brand Products
Click to view available options
Diameter (Metric) Bottom:
8 mm
10.5 mm
47 mm
Diameter (Metric) Top:
12 mm
14.5 mm
55 mm
Unit Size:
Pack of 10
Pack of 5
Product Code. 15231429
Brand: BRAND™ 144310
Only null left
Add to basket
Add to basket
Natural colored stopper is constructed of silicone.
  • Constructed of silicone
  • Natural-colored
TRUSTED_SUSTAINABILITY
Specifications
Height (Metric) 20 mm
Diameter (Metric) Bottom 8 mm
Diameter (Metric) Top 12 mm
Material Silicone
Product Type Silicone Stopper