Learn More
DWK Life Sciences Silicone sealing ring, for GL threads, without PTFE washer
Product Code. p-7189316 Shop All DWK Life Sciences Products
Click to view available options
Diameter (Metric) Inside:
6 mm
8 mm
10 mm
12 mm
14 mm
16 mm
18 mm
20 mm
26 mm
32 mm
Joint Size:
GL 14
GL 18
GL 25
GL 32
GL 45
Unit Size:
Each
Pack of 10
Product Code. 15931541
Brand: DWK Life Sciences 286200405
Seal for screw connection cap.
Suitable for PBT screw cap. Exemplary application: Fixing tubes
  • Suitable for PBT screw cap
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inside 6 mm
For Use With (Application) Fixing tubes; Sealing
Joint Size GL 14
Quantity 10/Pk.
Diameter (Metric) Outside 12 mm
Material Silicone