Learn More
DWK Life Sciences Silicone sealing ring, for GL threads, without PTFE washer
Product Code. 15369797 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric) Inside:
6 mm
8 mm
10 mm
12 mm
14 mm
16 mm
18 mm
20 mm
26 mm
32 mm
Joint Size:
GL 14
GL 18
GL 25
GL 32
GL 45
Unit Size:
Each
Pack of 10
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inside Joint Size Unit Size
15369797 8 mm GL 25 Pack of 10
15931541 6 mm GL 14 Pack of 10
10345191 6 mm GL 18 Each
15874598 10 mm GL 18 Pack of 10
10548411 26 mm GL 45 Each
15824618 14 mm GL 32 Pack of 10
10141831 8 mm GL 18 Each
15814618 12 mm GL 32 Pack of 10
10420303 10 mm GL 25 Each
15884598 12 mm GL 25 Pack of 10
15804618 10 mm GL 32 Pack of 10
15844618 18 mm GL 32 Pack of 10
15834618 16 mm GL 32 Pack of 10
15854618 20 mm GL 32 Pack of 10
15217458 32 mm GL 45 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 15369797 Supplier DWK Life Sciences Supplier No. 286201204
  • ONLINE EXCLUSIVE PRICE
    29.98€
    25.04€ / Pack of 10
    Save 4.94€
    16% Off
In Stock
Seal for screw connection cap.
Suitable for PBT screw cap. Exemplary application: Fixing tubes
  • Suitable for PBT screw cap
Specifications
Diameter (Metric) Inside 8 mm
For Use With (Application) Fixing tubes; Sealing
Joint Size GL 25
Quantity 10/Pk.
Diameter (Metric) Outside 22 mm
Material Silicone