missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hirschmann™ SILICOSEN Silicone Stoppers
Click to view available options
Color:
Green
Unit Size:
Pack of 10
Description
- Uniform pore structure
- Good air permeability
- Chemically and temperature resistant
- Liquid repellent and reusable
Specifications
Specifications
| Diameter (Metric) Inner | 24-33mm |
| For Use With (Application) | For The Preparation & Sterilization Of Culture Media |
| Length (Metric) | 20 mm |
| Quantity | 10/Pk. |
| Color | Green |
| Material | Silicone |
| Product Type | Plug |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction