Learn More
Ismatec™ Silicone Peroxide Peristaltic Tubing
Click to view available options
Diameter (Metric) Inner:
2 mm
Unit Size:
25m
Description
Order Info
DiameterMetricInside
Specifications
Specifications
| Diameter (Metric) Inner | 2 mm |
| Material | Silicone (Peroxide-cured) |
| For Use With | Low Pressure Pumps |
| Product Type | Peristaltic Tubing |