Learn More
Elma™ Silicone Holder Set
Description
- Easy handling and positioning
- For securing Elmasonic accessories
Specifications
Specifications
| Quantity | 10 Holders/Set |
| Type | Holder |
| Diameter (Metric) | 20 mm |
| For Use With (Equipment) | Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY |
| Height (Metric) | 20 mm |
| Material | Silicone |
| Weight (Metric) | 4 g |