Learn More
Borer Chemie™ Silicone-free Grease Tube
Product Code. p-8014373
Click to view available options
Type:
High-Vacuum
Standard
Unit Size:
60g
Each
Product Code. 10404681
Brand: Borer Chemie™ 514215.00CA15
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Standard quality for desiccators, ground joints, stopcocks and apparatuses. Borer Chemie™ Silicone-Free Grease Tubes are suitable for sealing and lubricating fixed and rotary ground glass joints and for greasing fittings made of plastics and metal.
  • Excellent thermal stability
  • Available in standard quality (vacuum tight up to 300°C) and high-vacuum quality (up to 400°C)
  • Constant lubricating consistency between -40°C and +250°C
  • No dripping point
  • Chemically stable against most acids, alkalis and gases
  • Insoluble in water and low molecular mono- and polyhydric alcohols
Specifications
Type Standard
Quantity 60 g