Learn More
Borer Chemie™ Silicone-free Grease Tube
Description
- Excellent thermal stability
- Available in standard quality (vacuum tight up to 300°C) and high-vacuum quality (up to 400°C)
- Constant lubricating consistency between -40°C and +250°C
- No dripping point
- Chemically stable against most acids, alkalis and gases
- Insoluble in water and low molecular mono- and polyhydric alcohols
Specifications
Specifications
| Product Type | High-Vacuum |
| Quantity | 60 g |