Learn More
Watson-Marlow Tubing Silicone Double Segment Manifold Tubing
Product Code. 17847973
Click to view available options
Diameter (Metric) Inner:
1.29 mm
2.29 mm
Unit Size:
Pack of 6
Product Code. 17847973
Brand: Watson-Marlow Tubing 983.0129.000
Only null left
Add to basket
Add to basket
Silicone Double Segment Manifold Tubing is pump tubing tor 401U/D1, 401U/DM2 and 401U/DM3 manual/auto control ultra compact pumps.
  • 401U/D1, 401U/DM2 and 401U/DM3 Manual/auto Control Ultra Compact Pumps
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Inner 0.05 in.
Diameter (Metric) Inner 1.29 mm
Material Silicone
Overall Length (English) 1.5 ft.
Overall Length (Metric) 0.46 m
Thickness (English) 0.03 in.
Thickness (Metric) 0.8 mm
Product Type Double Segment Tubing