Learn More
MP EcoChrom Silica DCC, 60 Å, 63 to 200 μm, pH 7.5, MP Biomedicals™
Product Code. 17877044 Shop All MP Biomedical Products
Change view
Click to view available options
Quantity:
3 kg
Unit Size:
3kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17877044 3 kg 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17877044 Supplier MP Biomedicals Supplier No. 0204526
Specifications
CAS 7631-86-9
Quantity 3 kg
Molecular Formula O2Si
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES O=[Si]=O
IUPAC Name silanedione
Molecular Weight (g/mol) 60.08
PubChem CID 24261
ChEBI CHEBI:30563
Formula Weight 60.08
Density 2g/cm3 at 20°C
Chemical Name or Material Silica, DCC