Learn More
Silica Active 36-200μm, 60Å, EcoChrom™, MP Biomedicals™
Product Code. 11313559 Shop All MP Biomedical Products
Click to view available options
Quantity:
500 g
Unit Size:
500g
Product Code. 11313559
Brand: MP Biomedicals 0202769
For Chromatography; Chemical Synthesis
TRUSTED_SUSTAINABILITY
Specifications
CAS 7631-86-9
Color White
Physical Form Powder
Quantity 500 g
Molecular Formula O2Si
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES O=[Si]=O
IUPAC Name dioxosilane
Molecular Weight (g/mol) 60.08
PubChem CID 24261
ChEBI CHEBI:30563
Formula Weight 60.08
Density 2g/cm3 at 20°C
Chemical Name or Material Silica, Active