Learn More
Tocris Bioscience™ Sildenafil citrate
Description
Orally active, potent inhibitor of phosphodiesterase 5 (PDE5) (IC50 = 4 nM). Enhances nitric oxide-dependent relaxation of human corpus cavernosum in vitro.
Specifications
Specifications
| Chemical Name or Material | 1-[[3-(4,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]-4-methylpiperazine citrate |
| CAS | 171599-83-0 |
| Assay Percent Range | >99% |
| Quantity | 25 mg |
| Molecular Formula | C22H30N6O4S.C6H8O7 |
| Formula Weight | Observed MW: 666.7 |