Learn More
BRAND™ SILBERBRAND Automatic Burette Spare Burette Tube
Shop All Brand Products
Click to view available options
Capacity (Metric):
25 mL
50 mL
Color:
Amber
Clear
Unit Size:
Each
Description
- For use with automatic burette
- Dr. Scilling pattern
Specifications
Specifications
| Description | Spare burette f. aut.burette Schilling 25 ml SILBERBRAND Schellb., AR-GLAS™, amber glass, l. 390 mm |
| Color | Amber |
| Capacity (Metric) | 25 mL |
| Length (Metric) | 390 mm |
| For Use With (Equipment) | Automatic Burette |
| Material | Borosilicate Glass (3.3) |
| Type | Spare Buret |