Learn More
Epredia™ Signature Series™ Flex™ Dehydrating Reagent
Product Code. 12638106 Shop All Epredia Products
Click to view available options
Quantity:
1 gal.
Product Type:
Flex 100 Dehydrating Reagent
Flex 80 Dehydrating Reagent
Flex 95 Dehydrating Reagent
missing translation for 'unitSize'
Pack of 4
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 12638106
missing translation for 'mfr': Epredia™ 8301R
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Achieve brilliant cytoplasmic staining and complete dehydration with Epredia™ Signature Series Flex™ 100, 95, 80.
  • Patented formulation of methyl and isopropyl alcohol minimizes over-dehydration in smaller biopsies
  • Excellent for lipid extraction allowing for increased paraffin infiltration
  • May be used as a solvent for special stains
TRUSTED_SUSTAINABILITY
Specifications
Product Type Flex 80 Dehydrating Reagent
Quantity 1 gal.