Learn More
Epredia™ Signature Series Cover Glass
Product Code. 12658066 Shop All Epredia Products
Click to view available options
Dimensions (L x W):
40 x 24 mm
50 x 24 mm
60 x 24 mm
Unit Size:
Pack of 10
Product Code. 12658066
Brand: Epredia™ 12450S
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Eliminate sticking and reduce waste with the uniquely manufactured Epredia™ Signature Series Cover Glass. Ideal for automatic coverslippers.
Thickness 1:
  • 0.13 - 0.17mm
Ordering Information:
  • Case of 10 Pk. (68-72/pack)
  • Available in several sizes
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 24 mm
Width (Metric) 50 mm
Thickness (Metric) #1 (0.13 to 0.16 mm)
Dimensions (L x W) 50 x 24 mm
Quantity Case of 10 Pk. (68-72/pack)