Learn More
SymLine™ SignalBox
Click to view available options
Design:
1-channel
2-channel
5-channel
Plug Type:
EU, UK, US
EU-style
UK-style
US-style
Unit Size:
1 sheet
Each
Description
- Clever way to change a disposal container without interrupting the inflow.
- LEDs and acoustic buzzer warn you in due time before the filling level of your collection containers reaches a critical height.
- When container is changed, the alert can be muted via the new touchpad.
- Includes touchless alarm-resetting fields for up to five containers, up to five switch inputs and up to five switch outputs for peripheral units, and power supply cable with open end and clamp.
Spécification
Spécification
| Design | 2-channel |
| Type | Liquid Spill Alarm |
| For Use With (Equipment) | Safety Waste Cap and Electronic Level Controllers |
| Includes | SignalBox2, Power Supply Cable with Open End and Clamp |
| Dimensions (L x W x H) | 36 x 139 x 74 mm |
| Weight (English) | 1.32 lb. |
| Weight (Metric) | 0.6 kg |
| Plug Type | EU-style |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu