Learn More
SymLine™ Signal Boxes
Product Code. 15955801
Click to view available options
Length (Metric):
3 m
55 mm
86 mm
Product Type:
Signalbox T1
Signalbox T5
Plug Type:
EU-style
UK-style
US-style
Includes:
1 Sensor Input and 1 Potential-free Output
5 Sensor Inputs and 6 Potential-free Outputs
Alarm at Empty State
Alarm at Full State
T5 with 5 Disc Fill Level Sensors
Unit Size:
Each
Product Code. 15955801
Brand: SymLine™ 108122
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Monitor two filling levels at once. Ideal for integration into lab furniture with SymLine™ Built-in Signal Box.
  • Clever way to change disposal container without interrupting the inflow
  • LEDs and acoustic buzzer warn you before filling level of collection containers reaches critical height
  • When container is changed, alert can be muted via new touchpad
Specifications
Product Type Signalbox T1
For Use With (Equipment) SCAT™ Sensors and Safety Waste Caps
Length (English) 7.1 in.
Length (Metric) 55 mm
Plug Type UK-style
Size 180 x 105 x 55 mm
Voltage 24 V
Amperage 2 A
For Use With (Application) Monitoring Filling Levels
Height (Metric) 105 mm
Includes 1 Sensor Input and 1 Potential-free Output
Width (Metric) 180 mm