Learn More
Uvex™ Signal Waist Coat 8960-Yellow 100%PES
Product Code. p-7232010 Shop All Uvex Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Each
Product Code. 17256651
Brand: Uvex™ 8937310
Only null left
Add to basket
Add to basket
All that is essential is that everyone can be seen despite the poor visibility. The high-visibility clothing from the uvex protection flash collection uses bright reflective strips. And of course the cargo trousers and softshell jackets with their optimum fit can also be seen in daylight
  • Material: 100% polyester / mesh Safety vest made of polyester / mesh with space for logo attachment.
  • Velcro fastener, 2 circumferential reflective stripes in the torso area, reflective strips over the shoulders for better visibility in stooped position, edging edges, logo application possible (position: back)
  • Certified according to EN 20471 X2
  • Size: M (50/52) - XXXL (66/68)
  • Care instructions: 40° C washable, gentle cycle (max. 25 washes), chlorinate impossible, ironing impossible, dry cleaning: P (max. 20 cleaning cycles), tumble drying impossible
TRUSTED_SUSTAINABILITY
Specifications
Closure Type Hook and Loop
Product Type High-visibility Vest
Color Yellow
Size Medium
Material Polyester
Thickness (Metric) 140 g/m²